Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin 6 Antibody (RM0157-3L27), DyLight 680, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212423FR
Description
Cathepsin 6 Monoclonal specifically detects Cathepsin 6 in Mouse samples. It is validated for Western Blot.Specifications
Cathepsin 6 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Cts6 | |
Mouse recombinant Cathepsin 6 | |
0.1 mL | |
58518 | |
Store at 4C in the dark. | |
IgG2 |
Western Blot | |
RM0157-3L27 | |
Western Blot | |
1600022N02Rik | |
Rat | |
Protein A or G purified | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction