Learn More
Cathepsin 7 Antibody (RM0158-4A12), Alexa Fluor™ 750, Novus Biologicals™
SDP
Rat Monoclonal Antibody
Supplier:  Novus Biologicals NBP212424AF750
Catalog No. NP212424A70
Only null left
Add to Cart
Description
Description
Cathepsin 7 Monoclonal specifically detects Cathepsin 7 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Cathepsin 7
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
Cts7
Mouse recombinant Cathepsin 7
0.1 mL
56092
Store at 4°C in the dark.
Western Blot
RM0158-4A12
Western Blot
CTS1, Epcs24, Epcs71
Rat
Protein A or G purified
Primary
Mouse
IgG2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.