Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin 7 Antibody (RM0158-4A12), DyLight 650, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212424C
Description
Cathepsin 7 Monoclonal specifically detects Cathepsin 7 in Mouse samples. It is validated for Western Blot.Specifications
Cathepsin 7 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Cts7 | |
Mouse recombinant Cathepsin 7 | |
0.1 mL | |
56092 | |
Store at 4C in the dark. | |
IgG2 |
Western Blot | |
RM0158-4A12 | |
Western Blot | |
CTS1, Epcs24, Epcs71 | |
Rat | |
Protein A or G purified | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction