Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin A/Lysosomal Carboxypeptidase A Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298264R
Description
Cathepsin A/Lysosomal Carboxypeptidase A Polyclonal specifically detects Cathepsin A/Lysosomal Carboxypeptidase A in Mouse samples. It is validated for Western Blot, ELISA.Specifications
Cathepsin A/Lysosomal Carboxypeptidase A | |
Polyclonal | |
Western Blot, ELISA | |
beta-galactosidase 2, beta-galactosidase protective protein, Carboxypeptidase C, Carboxypeptidase L, cathepsin ANGBE, EC 3.4.16, EC 3.4.16.5, GLB2, GSL, lysosomal protective protein, PPCA, PPGBprotective protein for beta-galactosidase (galactosialidosis), Protective protein cathepsin A, Protective protein for beta-galactosidase | |
Produced in rabbits immunized with purified, recombinant Cathepsin A/Lysosomal Carboxypeptidase A (P16675-1; Met1-Tyr474). | |
0.1 mL | |
Signal Transduction | |
5476 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction