Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin A/Lysosomal Carboxypeptidase A Antibody (RM0160-4A12), Janelia Fluor™ 549, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP212071JF549
Description
Cathepsin A/Lysosomal Carboxypeptidase A Monoclonal specifically detects Cathepsin A/Lysosomal Carboxypeptidase A in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CathepsinA/LysosomalCarboxypeptidaseA | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CTSA | |
Mouse recombinant Cathepsin A/Lysosomal Carboxypeptidase A | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Frozen) | |
RM0160-4A12 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen | |
beta-galactosidase 2, beta-galactosidase protective protein, Carboxypeptidase C, Carboxypeptidase L, cathepsin ANGBE, EC 3.4.16, EC 3.4.16.5, GLB2, GSL, lysosomal protective protein, PPCA, PPGBprotective protein for beta-galactosidase (galactosialidosis), Protective protein cathepsin A, Protective protein for beta-galactosidase | |
Rat | |
Protein A or G purified | |
Signal Transduction | |
5476 | |
Store at 4°C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction