Learn More
Invitrogen™ Cathepsin A Recombinant Rabbit Monoclonal Antibody (104)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA529204
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human Cathepsin A/CTSA.
The cathepsin family of proteolytic enzymes include several diverse classes of proteases. Cathepsins B, L, H, K, S and O comprise the cysteine protease class. Cathepsins D and E comprise the aspartyle protease class. The serine protease class includes cathepsin G. Cathepsins function in cellular metabolism and participate in peptide biosynthesis and protein degradation. Cathepsin A, a serine carboxypeptidase, exists in a high molecular weight lysosomal complex with b-galactosidase (b-gal) and a-neuraminidase (Neu1). Cathepsin A functions to protect b-gal and Neu1 from intralysosomal proteolysis. Deficiencies in cathepsin A lead to deficiencies in b-gal and Neu1. The gene encoding human cathepsin A maps to chromosome 20q13.12. Mutations in this gene cause glactosialidosis, a lysosomal storage disorder resulting from the b-gal and Neu1 deficiencies.
Specifications
Cathepsin A | |
Recombinant Monoclonal | |
1.39 mg/mL | |
PBS with no preservative | |
P10619 | |
CTSA | |
Recombinant Human Cathepsin A/CTSA protein (Met1-Tyr480). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunocytochemistry | |
104 | |
Unconjugated | |
CTSA | |
AU019505; beta-galactosidase 2; beta-galactosidase protective protein; carboxypeptidase C; Carboxypeptidase L; carboxypeptidase Y-like kininase; carboxypeptidase-L; cathepsin A; CTSA; deamidase; GLB2; GSL; lysosomal carboxypeptidase A; lysosomal protective protein; Lysosomal protective protein 20 kDa chain; Lysosomal protective protein 32 kDa chain; NGBE; PPCA; PPGB; protective protein cathepsin A; Protective protein for beta-galactosidase; urinary kininase | |
Rabbit | |
Protein A | |
RUO | |
5476 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.