Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin B Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP119797
Description
Cathepsin B Polyclonal specifically detects Cathepsin B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Cathepsin B | |
Polyclonal | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
APP secretase, APPS, cathepsin B, Cathepsin B1, CPSBamyloid precursor protein secretase, cysteine protease, EC 3.4.22, EC 3.4.22.1 | |
Rabbit | |
Affinity Purified | |
RUO | |
1508 | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Western Blot 1.0 - 2.0 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:100, Immunohistochemistry-Paraffin 1:200 | |
P07858 | |
CTSB | |
Synthetic peptide made to an internal portion of the Cathepsin B human protein (between residues 200-270) [UniProt P07858] | |
0.1 mL | |
Primary | |
Immunogen has 84% homology to bovine and chicken. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction