Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cathepsin B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA580517
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Cathepsin B/CTSB.
Cathepsin B is a gene located on chromosome 8p23.1 that encodes a lysosomal cysteine protease involved in intracellular protein degradation. As a member of the cathepsin family, Cathepsin B plays a crucial role in the lysosomal pathway, facilitating the breakdown of proteins into amino acids, which can then be recycled for cellular functions. In addition to its proteolytic activity within lysosomes, Cathepsin B is also implicated in extracellular matrix remodeling, cellular invasion, and apoptotic pathways. Its expression and activity are often upregulated in various cancer types, including breast, colon, and pancreatic cancers, where it contributes to tumor progression and metastasis by degrading extracellular matrix components and promoting tumor cell invasion. Due to these roles, Cathepsin B is considered a potential biomarker for cancer prognosis and a target for therapeutic intervention aimed at inhibiting cancer cell dissemination. Research continues to explore its broader implications in diseases related to protease activity and inflammation.
Specifications
Cathepsin B | |
Polyclonal | |
Unconjugated | |
Ctsb | |
amyloid precursor protein secretase; APP secretase; APPS; cathepsin B; Cathepsin B heavy chain; Cathepsin B light chain; Cathepsin B1; CB; CPSB; Ctsb; cysteine protease; RSG-2 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
1508 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with no preservative | |
P07858 | |
Ctsb | |
Recombinant Human Cathepsin B/CTSB protein (Arg18-Ile339). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction