Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin B Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 122161AP150UL

Description
Cathepsin B is a lysosomal cysteine protease that has been implicated in the pathogenesis ofrhematoid arthritis, muscular dystrophy and tumour metastasis.Specifications
Cathepsin B | |
Polyclonal | |
Unconjugated | |
CTSB | |
APP secretase, APPS, cathepsin B, Cathepsin B1, CPSB, CTSB | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13030, 1508 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P07858, P10605 | |
Ctsb | |
Cathepsin B Fusion Protein Ag2898 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction