Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin D Antibody (010), mFluor Violet 500 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290119MFV500
Description
Cathepsin D Monoclonal antibody specifically detects Cathepsin D in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
Cathepsin D | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry (Paraffin) | |
010 | |
Western Blot, Immunohistochemistry-Paraffin | |
cathepsin D, cathepsin D (lysosomal aspartyl protease), CPSD, EC 3.4.23, EC 3.4.23.5, lysosomal aspartyl peptidase, lysosomal aspartyl protease, MGC2311, neuronal 10 | |
Rabbit immunized with purified, recombinant Human Cathepsin D (Uniprot#: P07339, Met1-Leu412) | |
0.1 mL | |
Cancer, Cellular Markers | |
1509 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction