Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin D Antibody (102), FITC, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290432F
Description
Cathepsin D Monoclonal specifically detects Cathepsin D in Mouse samples. It is validated for ELISA.Specifications
Cathepsin D | |
Monoclonal | |
FITC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
102 | |
ELISA | |
cathepsin D, cathepsin D (lysosomal aspartyl protease), CPSD, EC 3.4.23, EC 3.4.23.5, lysosomal aspartyl peptidase, lysosomal aspartyl protease, MGC2311, neuronal 10 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Cathepsin D (NP_034113.1; Met 1-Leu 410). | |
0.1 mL | |
Cancer, Cellular Markers | |
1509 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction