Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin E Antibody (215), DyLight 488, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290637G
Description
Cathepsin E Monoclonal specifically detects Cathepsin E in Mouse samples. It is validated for ELISA.Specifications
Cathepsin E | |
Monoclonal | |
DyLight 488 | |
CATE, cathepsin E, EC 3.4.23, EC 3.4.23.34, erythrocyte membrane aspartic proteinase, slow-moving proteinase | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Cathepsin E (NP_031825.2; Met1-Pro397). | |
0.1 mL | |
Adaptive Immunity, Immunology, Innate Immunity, Lipid and Metabolism | |
1510 | |
Mouse | |
Purified |
ELISA | |
215 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Mouse CTSB Mouse CTSC | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction