Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin E Antibody (215), DyLight 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290637DL594
Description
Cathepsin E Monoclonal specifically detects Cathepsin E in Mouse samples. It is validated for ELISA.Specifications
| Cathepsin E | |
| Monoclonal | |
| DyLight 594 | |
| CATE, cathepsin E, EC 3.4.23, EC 3.4.23.34, erythrocyte membrane aspartic proteinase, slow-moving proteinase | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Cathepsin E (NP_031825.2; Met1-Pro397). | |
| 0.1 mL | |
| Adaptive Immunity, Immunology, Innate Immunity, Lipid and Metabolism | |
| 1510 | |
| Mouse | |
| Purified |
| ELISA | |
| 215 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| No cross-reactivity in ELISA with: Mouse CTSB Mouse CTSC | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction