Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin E Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298452B
Description
Cathepsin E Polyclonal specifically detects Cathepsin E in Rat samples. It is validated for ELISA.Specifications
Cathepsin E | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Rat | |
Purified |
ELISA | |
Biotin | |
CATE, cathepsin E, EC 3.4.23, EC 3.4.23.34, erythrocyte membrane aspartic proteinase, slow-moving proteinase | |
Produced in rabbits immunized with purified, recombinant Cathepsin E (AAH62002.1; Met1-Pro398). | |
0.1 mL | |
Adaptive Immunity, Immunology, Innate Immunity, Lipid and Metabolism | |
1510 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction