Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin E Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$206.50 - $499.50
Specifications
Antigen | Cathepsin E |
---|---|
Dilution | Western Blot 1:2000, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:400 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB400152SS
![]() |
Novus Biologicals
NB400152SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB400152
![]() |
Novus Biologicals
NB400152 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
Cathepsin E Polyclonal specifically detects Cathepsin E in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Cathepsin E | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, Immunology, Innate Immunity, Lipid and Metabolism | |
Whole antisera with 0.05% Sodium Azide | |
1510 | |
A synthetic peptide made to the C-terminus of mouse Cathepsin E [UniProt# P70269] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot 1:2000, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:200, Immunohistochemistry-Paraffin 1:400 | |
Polyclonal | |
Antisera | |
RUO | |
Human, Mouse, Rat | |
CATE, cathepsin E, EC 3.4.23, EC 3.4.23.34, erythrocyte membrane aspartic proteinase, slow-moving proteinase | |
CTSE | |
IgG | |
Unpurified | |
42 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title