Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cathepsin E Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547076
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547076 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547076 Supplier Invitrogen™ Supplier No. PA547076
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody detects both pro and mature mouse Cathepsin E in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant human Cathepsin E and less than 1% cross-reactivity with recombinant mouse Cathepsin D is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Cathepsin E may have a role in immune function. It is most likely involved in the processing of antigenic peptides during MHC class II-mediated antigen presentation. It may also play a role in activation-induced lymphocyte depletion in the thymus, and in neuronal degeneration and glial cell activation in the brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin E
Applications Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene CTSE
Gene Accession No. P70269
Gene Alias A430072O03Rik; C920004C08Rik; CATE; Cathepsin E; Cathepsin E form I; Cathepsin E form II; CE; CEA; CEB; CTSE; Ctsea; EC 3.4.23.34; erythrocyte membrane aspartic proteinase; slow-moving proteinase
Gene Symbols CTSE
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin E Gln19-Pro397.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13034
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.