Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin E Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA316821
Description
Prior to immunostaining paraffin tissues, antigen retrieval with sodium citrate buffer (pH 6.0) is recommended. Suggested positive control: antigen standard for CTSE (transient overexpression lysate).
Cathepsin E may have a role in immune function. It is most likely involved in the processing of antigenic peptides during MHC class II-mediated antigen presentation. It may also play a role in activation-induced lymphocyte depletion in the thymus, and in neuronal degeneration and glial cell activation in the brain.
Specifications
Cathepsin E | |
Polyclonal | |
Unconjugated | |
CTSE | |
A430072O03Rik; C920004C08Rik; CATE; Cathepsin E; Cathepsin E form I; Cathepsin E form II; CE; CEA; CEB; CTSE; Ctsea; EC 3.4.23.34; erythrocyte membrane aspartic proteinase; slow-moving proteinase | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Conc. Not Determined | |
whole serum with 0.05% sodium azide | |
P14091, P16228, P70269 | |
CTSE | |
A synthetic peptide made to the C-terminus of mouse Cathepsin E. | |
RUO | |
13034, 1510, 25424 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction