Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin E Rabbit anti-Mouse, Clone: 215, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
| Antigen | Cathepsin E |
|---|---|
| Clone | 215 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
Description
Cathepsin E Monoclonal specifically detects Cathepsin E in Mouse samples. It is validated for ELISA.Specifications
| Cathepsin E | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| CATE, cathepsin E, EC 3.4.23, EC 3.4.23.34, erythrocyte membrane aspartic proteinase, slow-moving proteinase | |
| Recombinant Mouse Cathepsin E protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 215 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Adaptive Immunity, Immunology, Innate Immunity, Lipid and Metabolism | |
| 0.2 um filtered solution in PBS | |
| 1510 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title