Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cathepsin F Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587925
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587925 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587925 Supplier Invitrogen™ Supplier No. PA587925
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LAPPEWDWRS KGAVTKVKDQ GMCGSCWAFS VTGNVEGQWF LNQGTLLSLS EQELLDCDKM DKACMGGLPS NAYSAIKNLG GLETEDDYSY QGHMQSCNFS AEKAKVYIND SVELSQNEQK LAAWLAKRGP ISVAINAFGM QFYRHGISRP LRPLCSPWLI DHAVLLVGYG NRSDVPFWAI KNSWGTDWGE KGYYYLHRGS GACGVNTMAS SAVVD; Positive Samples: U-251MG, SKOV3; Cellular Location: Lysosome.

Cathepsins are papain family cysteine proteinases that represent a major component of the lysosomal proteolytic system. Cathepsins generally contain a signal sequence, followed by a propeptide and then a catalytically active mature region. The very long (251 amino acid residues) proregion of the cathepsin F precursor contains a C-terminal domain similar to the pro-segment of cathepsin L-like enzymes, a 50-residue flexible linker peptide, and an N-terminal domain predicted to adopt a cystatin-like fold. The cathepsin F proregion is unique within the papain family cysteine proteases in that it contains this additional N-terminal segment predicted to share structural similarities with cysteine protease inhibitors of the cystatin superfamily. This cystatin-like domain contains some of the elements known to be important for inhibitory activity. CTSF encodes a predicted protein of 484 amino acids which contains a 19 residue signal peptide. Cathepsin F contains five potential N-glycosylation sites, and it may be targeted to the endosomal/lysosomal compartment via the mannose 6-phosphate receptor pathway. The cathepsin F gene is ubiquitously expressed, and it maps to chromosome 11q13, close to the gene encoding cathepsin W.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin F
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.44 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Ctsf
Gene Accession No. Q9UBX1
Gene Alias AI481912; Cathepsin F; CATSF; CLN13; CTSF; HGNC:2531
Gene Symbols Ctsf
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 270-484 of human CTSF (NP_0037842).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8722
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.