Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cathepsin G Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579097
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat thymus tissue.
Cathepsin G is a serine protease found in azurophilic granules of neutrophils involved in phagocytic lysis of engulfed forein particles. It is also involved in NK cell cytotoxicity.
Specifications
Cathepsin G | |
Polyclonal | |
Unconjugated | |
CTSG | |
CATG; cathepsin G; CG; Ctsg; CTSG protein; MGC23078; Vimentin-specific protease; VSP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1511, 290257 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 5 mg BSA and 0.05 mg sodium azide | |
P08311, P17977 | |
CTSG | |
E.coli-derived human Cathepsin G recombinant protein (Position: A76-L255). | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction