Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cathepsin G Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589049
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589049 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589049 Supplier Invitrogen™ Supplier No. PA589049
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GAHNIQRREN TQQHITARRA IRHPQYNQRT IQNDIMLLQL SRRVRRNRNV NPVALPRAQE GLRPGTLCTV AGWGRVSMRR GTDTLREVQL RVQRDRQCLR IFGSYDPRRQ ICVGDR; Positive Samples: Mouse kidney, Rat kidney; Cellular Location: Cell surface.

Cathepsin G is a serine protease found in azurophilic granules of neutrophils involved in phagocytic lysis of engulfed forein particles. It is also involved in NK cell cytotoxicity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin G
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CTSG
Gene Accession No. P08311, P17977, P28293
Gene Alias CATG; cathepsin G; CG; Ctsg; CTSG protein; MGC23078; Vimentin-specific protease; VSP
Gene Symbols CTSG
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 75-190 of human Cathepsin G (NP_0019021).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13035, 1511, 290257
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.