Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cathepsin G Recombinant Rabbit Monoclonal Antibody (9K4D3)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA551214
Description
Immunogen sequence: TDTLREVQLR VQRDRQCLRI FGSYDPRRQI CVGDRRERKA AFKGDSGGPL LCNNVAHGIV SYGKSSGVPP EVFTRVSSFL PWIRTTMRSF KLLDQMETPL.
Cathepsin G is a serine protease found in azurophilic granules of neutrophils involved in phagocytic lysis of engulfed forein particles. It is also involved in NK cell cytotoxicity.
Specifications
Cathepsin G | |
Recombinant Monoclonal | |
1.5 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P08311 | |
CTSG | |
A synthetic peptide corresponding to a sequence within amino acids 156-255 of human Cathepsin G (NP_001902.1). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
ELISA, Immunocytochemistry | |
9K4D3 | |
Unconjugated | |
CTSG | |
CATG; cathepsin G; CG; Ctsg; CTSG protein; MGC23078; Vimentin-specific protease; VSP | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1511 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction