Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cathepsin H/K/L/V Recombinant Rabbit Monoclonal Antibody (2Q10F10)
GREENER_CHOICE

Catalog No. PIMA535614
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535614 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535614 Supplier Invitrogen™ Supplier No. MA535614
Add to Cart
Add to Cart

Cathepsin H/K/L/V Recombinant Rabbit Monoclonal Antibody (ARC1276)

Cathepsin H/K/L/V Recombinant Monoclonal Antibody for Western Blot

The protein encoded by this gene is a lysosomal cysteine proteinase that plays a major role in intracellular protein catabolism. Its substrates include collagen and elastin, as well as alpha-1 protease inhibitor, a major controlling element of neutrophil elastase activity. The encoded protein has been implicated in several pathologic processes, including myofibril necrosis in myopathies and in myocardial ischemia, and in the renal tubular response to proteinuria. This protein, which is a member of the peptidase C1 family, is a dimer composed of disulfide-linked heavy and light chains, both produced from a single protein precursor. Multiple alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin H/K/L/V
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 2Q10F10
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CTSH
Gene Accession No. O35186, O60911, P00786, P06797, P07154, P07711, P09668, P43235, P49935, P55097
Gene Alias ACC4; ACC-4; ACC5; ACC-5; AI323530; AL022844; aleurain; Cat H; Cat K; cathepsin B3; Cathepsin BA; cathepsin H; Cathepsin H heavy chain; Cathepsin H light chain; Cathepsin H mini chain; Cathepsin K; cathepsin L2; cathepsin L2, preproprotein; cathepsin O; cathepsin O1; cathepsin O2; Cathepsin U; cathepsin V; Cathepsin X; catK; CATL2; CPSB; CTS02; CTSH; Ctsk; CTSL2; CTSO; CTSO1; CTSO2; CTSU; CTSV; minisatellite 10q detected by probe MMS10; MMS10-Q; Ms10q; N-benzoylarginine-beta-naphthylamide hydrolase; PKND; Pro-cathepsin H; PYCD; UNQ268/PRO305
Gene Symbols CTSH, CTSK, CTSL, CTSV
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 50-150 of human Cathepsin L/V/K/H (P07711).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13036, 13038, 13039, 1512, 1513, 1514, 1515, 25425, 25697, 29175
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.