Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cathepsin H/K/L/V Recombinant Rabbit Monoclonal Antibody (JM10-78)
GREENER_CHOICE

Catalog No. PIMA532602
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532602 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532602 Supplier Invitrogen™ Supplier No. MA532602
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

The protein encoded by this gene is a lysosomal cysteine proteinase that plays a major role in intracellular protein catabolism. Its substrates include collagen and elastin, as well as alpha-1 protease inhibitor, a major controlling element of neutrophil elastase activity. The encoded protein has been implicated in several pathologic processes, including myofibril necrosis in myopathies and in myocardial ischemia, and in the renal tubular response to proteinuria. This protein, which is a member of the peptidase C1 family, is a dimer composed of disulfide-linked heavy and light chains, both produced from a single protein precursor. Multiple alternatively spliced transcript variants have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin H/K/L/V
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Recombinant Monoclonal
Clone JM10-78
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene CTSH
Gene Accession No. O35186, O60911, P00786, P06797, P07154, P07711, P09668, P43235, P49935, P55097
Gene Alias ACC4; ACC-4; ACC5; ACC-5; AI323530; AL022844; aleurain; Cat H; Cat K; cathepsin B3; Cathepsin BA; cathepsin H; Cathepsin H heavy chain; Cathepsin H light chain; Cathepsin H mini chain; Cathepsin K; cathepsin L2; cathepsin L2, preproprotein; cathepsin O; cathepsin O1; cathepsin O2; Cathepsin U; cathepsin V; Cathepsin X; catK; CATL2; CPSB; CTS02; CTSH; Ctsk; CTSL2; CTSO; CTSO1; CTSO2; CTSU; CTSV; minisatellite 10q detected by probe MMS10; MMS10-Q; Ms10q; N-benzoylarginine-beta-naphthylamide hydrolase; PKND; Pro-cathepsin H; PYCD; UNQ268/PRO305
Gene Symbols CTSH, CTSK, CTSL, CTSV
Host Species Rabbit
Immunogen Synthetic peptide within Human Cathepsin aa 92-138.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13036, 13038, 13039, 1512, 1513, 1514, 1515, 25425, 25697, 29175
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.