Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cathepsin H Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547041
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547041 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547041 Supplier Invitrogen™ Supplier No. PA547041
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human (rh) Cathepsin H is observed and less than 2% cross-reactivity with recombinant mouse (rm) Cathepsin B, rmCathepsin C, rmCathepsin Z, rmCathepsin L, and rhCathepsin O is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

The cathepsin family of proteolytic enzymes contains several diverse classes of proteases. The cysteine protease class comprises cathepsins B, L, H, K, S, and O. The aspartyl protease class is composed of cathepsins D and E. Cathepsin G is in the serine protease class. Most cathepsins are lysosomal and each is involved in various cellular events such as peptide biosynthesis and protein degradation. The propeptide region of cathepsin H is highly homologous to other cathepsins. This region is thought to play a role in the processing of cysteine proteases. Cathepsin H is thought to be involved in several pathological conditions, including neuro degenerative disorders and cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin H
Applications Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene CTSH
Gene Accession No. P49935
Gene Alias ACC4; ACC-4; ACC5; ACC-5; AL022844; aleurain; Cat H; CATH; cathepsin B3; Cathepsin BA; cathepsin H; Cathepsin H heavy chain; Cathepsin H light chain; Cathepsin H mini chain; CPSB; Ctsh; N-benzoylarginine-beta-naphthylamide hydrolase; pro-cathepsin H
Gene Symbols CTSH
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin H Glu22-Val333.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13036
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.