Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cathepsin H Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575439
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The cathepsin family of proteolytic enzymes contains several diverse classes of proteases. The cysteine protease class comprises cathepsins B, L, H, K, S, and O. The aspartyl protease class is composed of cathepsins D and E. Cathepsin G is in the serine protease class. Most cathepsins are lysosomal and each is involved in various cellular events such as peptide biosynthesis and protein degradation. The propeptide region of cathepsin H is highly homologous to other cathepsins. This region is thought to play a role in the processing of cysteine proteases. Cathepsin H is thought to be involved in several pathological conditions, including neuro degenerative disorders and cancer.
Specifications
Cathepsin H | |
Polyclonal | |
Unconjugated | |
CTSH | |
ACC4; ACC-4; ACC5; ACC-5; AL022844; aleurain; Cat H; CATH; cathepsin B3; Cathepsin BA; cathepsin H; Cathepsin H heavy chain; Cathepsin H light chain; Cathepsin H mini chain; CPSB; Ctsh; N-benzoylarginine-beta-naphthylamide hydrolase; pro-cathepsin H | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13036, 1512, 25425 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P00786, P09668, P49935 | |
CTSH | |
Synthetic peptide to amino acids 171-210 of Human Cathepsin H. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction