Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cathepsin H Recombinant Rabbit Monoclonal Antibody (JE56-25)
GREENER_CHOICE

Catalog No. PIMA547478
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547478 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547478 Supplier Invitrogen™ Supplier No. MA547478
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: PC-3M cell lysates, PANC-1. Subcellular Location: Lysosome. Predicted band size: 37 kDa.

The cathepsin family of proteolytic enzymes contains several diverse classes of proteases. The cysteine protease class comprises cathepsins B, L, H, K, S, and O. The aspartyl protease class is composed of cathepsins D and E. Cathepsin G is in the serine protease class. Most cathepsins are lysosomal and each is involved in various cellular events such as peptide biosynthesis and protein degradation. The propeptide region of cathepsin H is highly homologous to other cathepsins. This region is thought to play a role in the processing of cysteine proteases. Cathepsin H is thought to be involved in several pathological conditions, including neuro degenerative disorders and cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin H
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE56-25
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CTSH
Gene Accession No. P09668
Gene Alias ACC4; ACC-4; ACC5; ACC-5; AL022844; aleurain; Cat H; CATH; cathepsin B3; Cathepsin BA; cathepsin H; Cathepsin H heavy chain; Cathepsin H light chain; Cathepsin H mini chain; CPSB; Ctsh; N-benzoylarginine-beta-naphthylamide hydrolase; pro-cathepsin H
Gene Symbols CTSH
Host Species Rabbit
Immunogen Synthetic peptide within human Cathepsin H aa 180-230.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1512
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.