Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin K Antibody (CTSK/2791), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279889H
Description
Cathepsin K Monoclonal specifically detects Cathepsin K in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Cathepsin K | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
CTSK | |
Recombinant fragment of human Cathepsin K protein (around aa 163-274) (exact sequence is proprietary) (Uniprot: P43235) | |
0.1 mL | |
Cancer | |
Human | |
IgG1 κ |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
CTSK/2791 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
cathepsin K, cathepsin K (pycnodysostosis), Cathepsin O, cathepsin O1, Cathepsin O2, Cathepsin X, CTS02, CTSO, CTSO1, CTSO2, EC 3.4.22, EC 3.4.22.38, MGC23107, PKND, PYCD | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction