Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cathepsin K Monoclonal Antibody (C1), Invitrogen™

Catalog No. PIMA543914
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543914 20 μL
PIMA543913 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543914 Supplier Invitrogen™ Supplier No. MA543914
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The protein encoded by this gene is a lysosomal cysteine proteinase involved in bone remodeling and resorption. This protein, which is a member of the peptidase C1 protein family, is predominantly expressed in osteoclasts. However, the encoded protein is also expressed in a significant fraction of human breast cancers, where it could contribute to tumor invasiveness. Mutations in this gene are the cause of pycnodysostosis, an autosomal recessive disease characterized by osteosclerosis and short stature. This gene may be subject to RNA editing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin K
Applications Western Blot
Classification Monoclonal
Clone C1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene CTSK
Gene Accession No. O35186, P43235, P43236, P55097, Q3ZKN1, Q5E968, Q9GLE3
Gene Alias AI323530; Cat K; cathepsin K; Cathepsin O; cathepsin O1; cathepsin O2; Cathepsin X; catK; CTS02; CTSK; CTSO; CTSO1; CTSO2; MGC23107; minisatellite 10q detected by probe MMS10; MMS10-Q; Ms10q; PKND; PYCD
Gene Symbols CTSK
Host Species Mouse
Immunogen Ala115-Met329 (Accession P43235), with N-terminal His Tag.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009334, 100054991, 101108972, 13038, 1513, 29175, 397569, 513038, 608843
Target Species Human, Mouse, Rat, Canine, Rabbit, Bovine, Sheep, Pig, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.