Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin K, Mouse anti-Human, Clone: CTSK/2791, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279754100UG
Description
Cathepsin K Monoclonal specifically detects Cathepsin K in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Cathepsin K | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| P43235 | |
| CTSK | |
| Recombinant fragment of human Cathepsin K protein (around aa 163-274) (exact sequence is proprietary) | |
| 100 μg | |
| Cancer | |
| 1513 | |
| Store at 4°C. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| CTSK/2791 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| cathepsin K, cathepsin K (pycnodysostosis), Cathepsin O, cathepsin O1, Cathepsin O2, Cathepsin X, CTS02, CTSO, CTSO1, CTSO2, EC 3.4.22, EC 3.4.22.38, MGC23107, PKND, PYCD | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction