Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin L Antibody (002), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289527AF647
Description
Cathepsin L Monoclonal antibody specifically detects Cathepsin L in Human samples. It is validated for ELISASpecifications
Cathepsin L | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
002 | |
ELISA | |
cathepsin L, cathepsin L1, CATL, CTSLEC 3.4.22.15, EC 3.4.22, FLJ31037, Major excreted protein, MEP | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Cathepsin L (NP_001903.1; Met1-Val333). | |
0.1 mL | |
Cancer, Cellular Markers | |
1514 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction