Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin L Antibody (33/2), Novus Biologicals™
SDP

Catalog No. NB1001775 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001775 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001775 Supplier Novus Biologicals Supplier No. NB1001775
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 6 publications

Cathepsin L Monoclonal specifically detects Cathepsin L in Human, Mouse, Rat, Mustelid samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Microarray.

Specifications

Antigen Cathepsin L
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 33/2
Conjugate Unconjugated
Dilution Western Blot 1:200, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen 1:10-1:500, Microarray
Formulation Ascites with 15mM Sodium Azide
Gene Accession No. P07711
Gene Alias cathepsin L, cathepsin L1, CATL, CTSLEC 3.4.22.15, EC 3.4.22, FLJ31037, Major excreted protein, MEP
Gene Symbols CTSL
Host Species Mouse
Immunogen Purified full length native protein (Human lung cancer line EPLC 32 M1).
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1514
Test Specificity Reacts specifically with native and denatured forms of human cathepsin L and procathepsin L. Recognizes an epitope within amino acid residues GYGFEST (265-271 in procathepsin L and 169-175 in the mature cathepsin L molecule). The antibody may be used for the suppression of the malignant growth of myeloma cells. Cross-reactivity is observed with rat cathepsin L (strong), mouse procathepsin L (weak) and mink cathepsin and procathepsin L. Does not cross-react with human cathepsin types B, D, H and S, procathepsins B and D, rat cathepsin B, and mouse procathepsins B and D.
Target Species Human, Mouse, Rat, Mustelid
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.