Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$330.00 - $547.00
Specifications
Antigen | Cathepsin L |
---|---|
Dilution | Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Cathepsin L Polyclonal antibody specifically detects Cathepsin L in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Cathepsin L | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Cancer, Cellular Markers | |
PBS, pH 7.2, 40% glycerol | |
1514 | |
IgG | |
Affinity purified |
Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human | |
cathepsin L, cathepsin L1, CATL, CTSLEC 3.4.22.15, EC 3.4.22, FLJ31037, Major excreted protein, MEP | |
This antibody was developed against Recombinant Protein corresponding to amino acids: GIASATLTFDHSLEAQWTKWKAMHNRLYGM | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title