Learn More
Invitrogen™ Cathepsin L Monoclonal Antibody (33/1), FITC, eBioscience™

Description
Cathepsin L is a lysosomal cysteine proteinase consisting of a heavy chain of about 25 kDa and a light chain of about 5 kDa derived proteolytically from the same precursor. Cathepsin L has been shown to be produced by many different cell types. Since cathepsin L is capable of degrading protein constituents of the extracellular matrix it is assumed to play a crucial role in tumor progression and metastasis and a number of other disorders where the destruction of the extracellular matrix is the major cause of disease (e.g. rheumatoid arthritis, neurodegeneration). Purity: >95%. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.
Specifications
Specifications
| Antigen | Cathepsin L |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 33/1 |
| Concentration | 10 μL/Test |
| Conjugate | FITC |
| Formulation | TBS with 1% BSA and 0.02% sodium azide; pH 7.2-7.4 |
| Gene | CTSL |
| Gene Accession No. | P07711 |
| Gene Alias | 1190035F06Rik; Cat L; cathepsin L; Cathepsin L1; Cathepsin L1 heavy chain; Cathepsin L1 light chain; CATHL; CatL; CP-2; Ctsl; CTSL1; cyclic protein 2; fs; furless; major excreted protein; MEP; nackt; nkt; p39 cysteine proteinase; Procathepsin L |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.