Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin L Rabbit anti-Human, Clone: 221, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
| Antigen | Cathepsin L |
|---|---|
| Clone | 221 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Cathepsin L Monoclonal specifically detects Cathepsin L in Human samples. It is validated for Western Blot.Specifications
| Cathepsin L | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| cathepsin L, cathepsin L1, CATL, CTSLEC 3.4.22.15, EC 3.4.22, FLJ31037, Major excreted protein, MEP | |
| Recombinant Human Cathepsin L protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 221 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cellular Markers | |
| 0.2 um filtered solution in PBS | |
| 1514 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title