Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin L/V/K/H Antibody (SR1633), Novus Biologicals™
SDP

Catalog No. NB258834 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB258834 100 μg
NB259872 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB258834 Supplier Novus Biologicals Supplier No. NBP321720100UL
Only null left
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Cathepsin L/V/K/H Monoclonal antibody specifically detects Cathepsin L/V/K/H in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence

Specifications

Antigen Cathepsin L/V/K/H
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone SR1633
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias ACC4, ACC-4, ACC5, ACC-5, aleurain, cathepsin B3, cathepsin BA, cathepsin K, cathepsin L1, cathepsin L2, cathepsin L2, preproprotein, Cathepsin O, cathepsin O1, Cathepsin O2, Cathepsin U, Cathepsin X, CATL, CATL2, CPSB, CTS02, CTSL1, CTSL2, CTSO, CTSO1, CTSO2, CTSU, Major excreted protein, MEP, N-benzoylarginine-beta-naphthylamide hydrolase, PKND, procathepsin L, PYCD
Host Species Rabbit
Immunogen A synthesized peptide derived from human Cathepsin L/V/K/H (Uniprot #: P07711, O60911, P43235, P09668)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1514
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.