Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin L/V/K/H Rabbit anti-Human, Mouse, Rat, Clone: 2Q10F10, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP316524100UL
This item is not returnable.
View return policy
Description
Cathepsin L/V/K/H Monoclonal antibody specifically detects Cathepsin L/V/K/H in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Cathepsin L/V/K/H | |
Monoclonal | |
Unconjugated | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
2Q10F10 | |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
ACC4, ACC-4, ACC5, ACC-5, aleurain, cathepsin B3, cathepsin BA, cathepsin K, cathepsin L1, cathepsin L2, cathepsin L2, preproprotein, Cathepsin O, cathepsin O1, Cathepsin O2, Cathepsin U, Cathepsin X, CATL, CATL2, CPSB, CTS02, CTSL1, CTSL2, CTSO, CTSO1, CTSO2, CTSU, Major excreted protein, MEP, N-benzoylarginine-beta-naphthylamide hydrolase, PKND, procathepsin L, PYCD | |
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human Cathepsin L/V/K/H (P07711). | |
100 μg | |
Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
1514 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction