Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin O Antibody (MM0131-9G31), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212083AF594
Description
Cathepsin O Monoclonal specifically detects Cathepsin O in Human samples. It is validated for Western Blot.Specifications
Cathepsin O | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
CTSO | |
Human recombinant Cathepsin O | |
Primary | |
Human | |
IgG2 |
Western Blot | |
MM0131-9G31 | |
Western Blot | |
cathepsin O, CTSO1, EC 3.4.22.42 | |
Mouse | |
0.1 mL | |
1519 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction