Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin O Antibody (MM0131-9G31), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212083MFV610
Description
Cathepsin O Monoclonal antibody specifically detects Cathepsin O in Human samples. It is validated for Western BlotSpecifications
Cathepsin O | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
1519 | |
Store at 4C in the dark. | |
IgG2 |
Western Blot | |
MM0131-9G31 | |
Western Blot | |
cathepsin O, CTSO1, EC 3.4.22.42 | |
Human recombinant Cathepsin O | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction