Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin S Antibody (054), PE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290713PE
Description
Cathepsin S Monoclonal antibody specifically detects Cathepsin S in Mouse samples. It is validated for ELISASpecifications
Cathepsin S | |
Monoclonal | |
PE | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
054 | |
ELISA | |
cathepsin S, EC 3.4.22, EC 3.4.22.27, FLJ50259, MGC3886 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Cathepsin S (Accession#: AAB94925.1; Met1-Ile340). | |
0.1 mL | |
Immunology | |
1520 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction