Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin S Antibody (9B10), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Cathepsin S |
---|---|
Clone | 9B10 |
Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
Applications | ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
Cathepsin S Monoclonal antibody specifically detects Cathepsin S in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
Cathepsin S | |
ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
cathepsin S, EC 3.4.22, EC 3.4.22.27, FLJ50259, MGC3886 | |
A synthesized peptide derived from Human Cathepsin S [UniProt P25774] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
9B10 | |
ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Immunology | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
1520 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title