Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin S Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABC503MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC503MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC503MI Supplier MilliporeSigma Supplier No. ABC503
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Cathepsin S. is a rabbit polyclonal antibody that detects Cathepsin S and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin S
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols CTSS
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 21 amino acids from the internal region of human cathepsin S.
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_004070.3.
Test Specificity This polyclonal antibody detects cathepsin S in human cells. It targets an epitope within 21 amino acids from the internal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.