Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Cathepsin S Recombinant Rabbit Monoclonal Antibody (9B10)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA549897
Description
Cathepsin S Recombinant Monoclonal Antibody for IHC (P), ELISA
The protein CTSS, a member of the peptidase C1 family, is a lysosomal cysteine proteinase that may participate in the degradation of antigenic proteins to peptides for presentation on MHC class II molecules. The protein can function as an elastase over a broad pH range in alveolar macrophages.
Specifications
Cathepsin S | |
Recombinant Monoclonal | |
0.8 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P25774 | |
CTSS | |
A synthesized peptide derived from human Cathepsin S. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
9B10 | |
Unconjugated | |
CTSS | |
Cat S; cathepsin S; Cats; CTSS | |
Rabbit | |
Affinity Chromatography | |
RUO | |
1520 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction