Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin V Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP326082MFV610
Description
Cathepsin V Polyclonal antibody specifically detects Cathepsin V in Human samples. It is validated for ELISASpecifications
Cathepsin V | |
Polyclonal | |
ELISA | |
cathepsin L2, Cathepsin U, Cathepsin V, CATL2, CTSUcathepsin L2, preproprotein, CTSVMGC125957, EC 3.4.22, EC 3.4.22.43 | |
Rabbits immunized with purified, recombinant Human Cathepsin V (Accession#: NP_001324.2, Met1-Val334) | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
1515 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction