Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cathepsin V Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547061
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547061 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547061 Supplier Invitrogen™ Supplier No. PA547061
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs, approximately 15% cross-reactivity with recombinant human (rh) Cathepsin C is observed and less than 5% cross-reactivity with rhCathepsin A, rhCathepsin D, rhCathepsin X/Z/P, rhCathepsin S, and rhCathepsin L is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

Cathepsin V (CTSU, CTSV, CATL2, cathepsin U, cathepsin L2) is a lysosomal cysteine proteinase that influences corneal physiology, and mediates degradation of invariant chain in human thymus. Cathepsin V is present in corneal epithelium, activated macrophages, and colorectal and breast carcinomas. A 1.8-kb mRNA to Cathepsin V is present in normal human thymus and testis. Cathepsin V is a member of the peptidase C1 family. Cysteine proteinases are synthesized as preproenzymes, which are processed to the corresponding proenzymes. The proenzymes are either targeted to the lysosome or continue along the cellular secretory route.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cathepsin V
Applications Immunohistochemistry (Paraffin), Neutralization, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene CTSV
Gene Accession No. O60911
Gene Alias Cathepsin L2; cathepsin L2, preproprotein; cathepsin U; Cathepsin V; CATL2; CTSL2; CTSU; CTSV; PRO305; UNQ268; UNQ268/PRO305
Gene Symbols CTSV
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Cathepsin V Val18-Val334.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1515
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.