Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin X/Z/P Antibody (009), DyLight 550, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290411R
Description
Cathepsin X/Z/P Monoclonal specifically detects Cathepsin X/Z/P in Mouse samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Cathepsin X/Z/P | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Cathepsin X/Z/P (NP_071720.1; Met 1-Val 306). | |
0.1 mL | |
Cancer, Epitope Tags | |
1522 | |
Mouse | |
Purified |
ELISA, Immunofluorescence | |
9 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human CTSZ | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction