Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin X/Z/P Antibody (6G11), Alexa Fluor™ 350, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP231211AF350

Catalog No. NP231211A30


Only null left
Add to Cart

Description

Description

Cathepsin X/Z/P Monoclonal specifically detects Cathepsin X/Z/P in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Cathepsin X/Z/P
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
Mouse
0.1 mL
Primary
Human
IgG2b κ
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
6G11
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
CTSZ
Recombinant human CTSZ (62-303aa) purified from E. coli
Cancer, Epitope Tags
1522
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.