Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin Z Antibody (05), DyLight 755 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP305906IR
Description
Cathepsin Z Monoclonal antibody specifically detects Cathepsin Z in Human samples. It is validated for ELISASpecifications
Cathepsin Z | |
Monoclonal | |
DyLight 755 | |
Cathepsin P, Cathepsin X, cathepsin Z, CTSX, EC 3.4.18.1, FLJ17088, preprocathepsin P | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Cathepsin Z (Q9UBR2; Met1-Val303). | |
0.1 mL | |
Cancer | |
1522 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
05 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction