Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin Z Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306499MFV610
Description
Cathepsin Z Polyclonal antibody specifically detects Cathepsin Z in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
Cathepsin Z | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence | |
Cathepsin P, Cathepsin X, cathepsin Z, CTSX, EC 3.4.18.1, FLJ17088, preprocathepsin P | |
Produced in rabbits immunized with E. coli-derived Human Cathepsin Z fragment. | |
0.1 mL | |
Cancer | |
1522 | |
Store at 4C in the dark. | |
IgG |
Immunocytochemistry | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction