Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cathepsin Z Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP305902PE
Description
Cathepsin Z Polyclonal antibody specifically detects Cathepsin Z in Mouse samples. It is validated for ELISASpecifications
Cathepsin Z | |
Polyclonal | |
ELISA | |
Cathepsin P, Cathepsin X, cathepsin Z, CTSX, EC 3.4.18.1, FLJ17088, preprocathepsin P | |
Produced in rabbits immunized with purified, recombinant Mouse Cathepsin Z (Accession#: NP_071720.1; Met1-Val306) | |
0.1 mL | |
Cancer | |
1522 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction